Learn More
MSC Dynalon Perforated Stir Blade, HDPE, FDA Compliant, Non-Sterile
Catalog No. 502836646
Encompass
This perforated stir blade is constructed from durable HDPE, offering reliability for various liquid handling and mixing applications. It is compliant with FDA standards.
  • Made from HDPE material.
  • FDA compliant resin/materials.
  • Non-sterile.
  • Not autoclavable.
  • Overall length of 22.23 cm.
  • Overall width of 8.9 cm.
  • Threaded components are interchangeable for a versatile and modular system.
Catalog No. 50-283-6646 Supplier MSC Supplier No. 35649763
This perforated stir blade is constructed from durable HDPE, offering reliability for various liquid handling and mixing applications. It is compliant with FDA standards.
  • Made from HDPE material.
  • FDA compliant resin/materials.
  • Non-sterile.
  • Not autoclavable.
  • Overall length of 22.23 cm.
  • Overall width of 8.9 cm.
  • Threaded components are interchangeable for a versatile and modular system.