Learn More
Sklar Instruments Perforated Tray, Sklar, Stainless Steel. Sklar's High Sided Perforated Trays are used for steam sterilization without moisture build up. The high sides are to protect the instruments during procedures. EA/1.
SDP
Catalog No. 501437142 Shop All Sklar Instruments Products
Encompass_Preferred
Perforated Tray, Sklar, Stainless Steel. Sklar's High Sided Perforated Trays are used for steam sterilization without moisture build up. The high sides are to protect the instruments during procedures. The products are made with a corrosion resistant surface and comes in a variety of sizes. This tray is 10 inches x 6-1/2 inches x 2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7142 Supplier Sklar Instruments Supplier No. 101874
Perforated Tray, Sklar, Stainless Steel. Sklar's High Sided Perforated Trays are used for steam sterilization without moisture build up. The high sides are to protect the instruments during procedures. The products are made with a corrosion resistant surface and comes in a variety of sizes. This tray is 10 inches x 6-1/2 inches x 2 inches.