Learn More
Enzo Life Sciences Perforin (mouse) monoclonal antibody (CB5.4) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7080100
Encompass_Preferred
Clone: CB5.4. Isotype: IgG2a. Immunogen: Recombinant mouse perforin (aa 98-534). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Mouse. Specificity: Recognizes an epitope in the region aa 402-534. Applications: ICC, IHC (FS), IHC (PS), IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-3246