Learn More
Edge Biosystems Inc Performa® DTR Gel Filtration Cartridges 108 pack
SDP
Catalog No. nc9701718
Encompass
The gold standard in gel filtration based dye terminator removal!Performa® DTR utilizes a fully hydrated gel filtration matrix that provides for a convenient and simple method of dye terminator removal for sequencing cleanup. Performa DTR ensures removal of BigDye® Terminator v3.1 or other dye terminators, along with dNTPs, salts and other low molecular weight materials from sequencing reactions. DNA primers and fragments up to 20 bases, buffers, nucleotides labeled with biotin, isotopes and other assorted markers are also eliminated. To minimize the potential for interference with sequencing applications, no preservatives, salts, or buffers are used in the preparation of Performa DTR.
Catalog No. NC9701718 Supplier Edge Biosystems Inc Supplier No. 42453
The gold standard in gel filtration based dye terminator removal!Performa® DTR utilizes a fully hydrated gel filtration matrix that provides for a convenient and simple method of dye terminator removal for sequencing cleanup. Performa DTR ensures removal of BigDye® Terminator v3.1 or other dye terminators, along with dNTPs, salts and other low molecular weight materials from sequencing reactions. DNA primers and fragments up to 20 bases, buffers, nucleotides labeled with biotin, isotopes and other assorted markers are also eliminated. To minimize the potential for interference with sequencing applications, no preservatives, salts, or buffers are used in the preparation of Performa DTR.