Learn More
Edge Biosystems Inc PERFORMA DTR CARTRIDGES PK/36
SDP
Catalog No. NC9294916
Encompass
fully hydrated gel filtration matrix that provides for a convenient and simple method of dye terminator removal for sequencing cleanup.
Catalog No. NC9294916 Supplier Edge Biosystems Inc Supplier No. 98780
fully hydrated gel filtration matrix that provides for a convenient and simple method of dye terminator removal for sequencing cleanup.