Learn More
THERMCO PRODUCTS Performa-Therm™ 10 L Validated Medical Transport Carrier
Description
- Maintains integrity, ensures compliance, and delivers life-saving treatments with temperature precision
- Ideal for short-term storage, transport, and delivery of temperature-sensitive materials
- Durable, reusable, washable nylon shell and molded tub with six PCM panels
- Validated for reliable temperature-controlled transport performance
- Certificate included
Specifications
Specifications
| Product Type | Medical Transport Tote |
| Format | Carrier |
| Sterility | Non-sterile |
| Closure Type | Zipper |
| Color | Blue |
| Includes | Molded Plastic Insert and 6 PCM Panels |
| Transport Temperature | Frozen |
| Volume (Metric) | 10 L |