Learn More
Thermco Products Performa-Therm™ PCM Panels, Frozen (-20™C)
Catalog No. 04500372
Change view
Click to view available options
Quantity:
Set of 6
Catalog No. Quantity
04-500-372 Set of 6
1 options
Catalog No. 04-500-372 Supplier Thermco Products Supplier No. ACCPTFRZ
Add to Cart
Add to Cart
Performa-Therm™ Phase Change Material (PCM) when frozen will maintain stable Frozen Temperature (-15°C to -25°C) when inserted into a Performa-Therm™ Medical Transport Carrier.
  • When frozen will maintain stable Frozen Temperature (-15°C to -25°C) when inserted into a Performa-Therm™ Medical Transport Carrier
Specifications
Product Type PCM Panels
Format Tube
Sterility Non-sterile
Color Pink
Quantity Set of 6
Material Plastic