Learn More
Edge Biosystems Inc PERFORMA V3
SDP
Supplier:  Edge Biosystems Inc 89939
Encompass
Performa DTR V3; 96-Well Short (10/Plates); Purifications: 960; Gel filtration plates consist of 440uL volume columns in a standardized array; Provides optimal performance for removal of BigDye; v3 and v3.1; Storage: 4 deg.C
Catalog No. NC9168446