Learn More
Grainger Regal Cutting Tools Taper Pipe Tap, 1/8-27 NPTF, Super Premium Steel, 1 ea
Catalog No. 502759643
Encompass_Preferred
This taper pipe tap is designed to meet standard requirements in the power generation industry. It is engineered for precision threading applications in materials such as stainless steel and aluminum.
  • Manufactured to Table 302 standards
  • Free cutting for quality threads
  • Extra long style
  • Bright finish
  • 1/8-27 NPTF thread size
  • H2 limit
Catalog No. 50-275-9643 Supplier Grainger Supplier No. 163ZV7
This taper pipe tap is designed to meet standard requirements in the power generation industry. It is engineered for precision threading applications in materials such as stainless steel and aluminum.
  • Manufactured to Table 302 standards
  • Free cutting for quality threads
  • Extra long style
  • Bright finish
  • 1/8-27 NPTF thread size
  • H2 limit