Learn More
Accuflow Systems Saint-Gobain Chemware Griffin Beaker, PTFE, 50 mL, Inert
Catalog No. NC3617491
Encompass_Preferred
Saint-Gobain Chemware PTFE Griffin beaker, 50 mL, provides chemically inert, reusable labware for handling aggressive reagents and ultra-pure processes. The fluoropolymer construction minimizes contamination risk and is suitable for applications where glass or other plastics are incompatible.
  • PTFE construction provides broad chemical resistance.
  • 50 mL capacity.
  • Griffin beaker shape for easy pouring and mixing.
  • Non-wetting surface reduces carryover and contamination.
  • Reusable, unbreakable, and heat resistant for routine lab use.
  • Suitable for aggressive reagent handling and ultra-pure processing.
Catalog No. NC3617491 Supplier Accuflow Systems Supplier No. D1069036
Saint-Gobain Chemware PTFE Griffin beaker, 50 mL, provides chemically inert, reusable labware for handling aggressive reagents and ultra-pure processes. The fluoropolymer construction minimizes contamination risk and is suitable for applications where glass or other plastics are incompatible.
  • PTFE construction provides broad chemical resistance.
  • 50 mL capacity.
  • Griffin beaker shape for easy pouring and mixing.
  • Non-wetting surface reduces carryover and contamination.
  • Reusable, unbreakable, and heat resistant for routine lab use.
  • Suitable for aggressive reagent handling and ultra-pure processing.