Learn More
Waters Corp Waters Check Valve, PerformancePLUS, HPLC Compatible, 2/pk
Catalog No. 50792726 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The PerformancePLUS™ Check Valve Cartridges are designed for pumps with the PerformancePLUS Cartridge Check Valve System. They function as inlet or outlet valves. Materials are selected for compatibility with all HPLC mobile phases, ensuring reliable performance. A two-piece housing allows easy cartridge replacement without disturbing the fitting, simplifying maintenance.
  • Designed for pumps with PerformancePLUS Cartridge Check Valve System.
  • Functions as either an inlet or outlet valve.
  • Materials compatible with all HPLC mobile phases.
  • Two-piece housing for easy cartridge replacement.
Catalog No. 50-792-726 Supplier Waters Corp Supplier No. 700002275
The PerformancePLUS™ Check Valve Cartridges are designed for pumps with the PerformancePLUS Cartridge Check Valve System. They function as inlet or outlet valves. Materials are selected for compatibility with all HPLC mobile phases, ensuring reliable performance. A two-piece housing allows easy cartridge replacement without disturbing the fitting, simplifying maintenance.
  • Designed for pumps with PerformancePLUS Cartridge Check Valve System.
  • Functions as either an inlet or outlet valve.
  • Materials compatible with all HPLC mobile phases.
  • Two-piece housing for easy cartridge replacement.