Learn More
Waters Corp Waters Corporation Vacuum Degasser Chamber, Stented, Performance Plus, for Alliance HPLC System
Catalog No. 502039870 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Plus Vacuum Degasser Chamber is a stented replacement component designed for Alliance HPLC Systems, serving as the standard degasser chamber for the Alliance 2695.
  • Designed as a replacement component
  • Features a stented degasser chamber
  • For use with Alliance HPLC Systems
  • Serves as the standard degasser chamber for Alliance 2695
  • Compatible with 2695 separations module
  • Compatible with 2695 D dissolution separations module
  • Compatible with 2795 separations module
  • Compatible with Waters InLine Degasser AF
Catalog No. 50-203-9870 Supplier Waters Corp Supplier No. 700011709
This Performance Plus Vacuum Degasser Chamber is a stented replacement component designed for Alliance HPLC Systems, serving as the standard degasser chamber for the Alliance 2695.
  • Designed as a replacement component
  • Features a stented degasser chamber
  • For use with Alliance HPLC Systems
  • Serves as the standard degasser chamber for Alliance 2695
  • Compatible with 2695 separations module
  • Compatible with 2695 D dissolution separations module
  • Compatible with 2795 separations module
  • Compatible with Waters InLine Degasser AF