Learn More
Waters Corp Check Valve Cartridge, PerformancePlus, Ceramic Ball & Seat, 2 Pack
Catalog No. 502039437 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The PerformancePlus™ Inlet Check Valve Cartridge is a high-quality replacement part designed for HPLC systems. Featuring a ceramic ball and seat, it helps to prevent sticking issues and is a direct drop-in replacement that requires no modifications. It is manufactured to ISO 9001 quality standards.
  • Designed to help with sticking issues
  • Direct drop-in replacement, requiring no modifications
  • Manufactured to ISO 9001 quality standards
  • Supplied as a two-pack
  • Features a ceramic ball and seat
  • Compatible with HPLC systems
Catalog No. 50-203-9437 Supplier Waters Corp Supplier No. 700002399
The PerformancePlus™ Inlet Check Valve Cartridge is a high-quality replacement part designed for HPLC systems. Featuring a ceramic ball and seat, it helps to prevent sticking issues and is a direct drop-in replacement that requires no modifications. It is manufactured to ISO 9001 quality standards.
  • Designed to help with sticking issues
  • Direct drop-in replacement, requiring no modifications
  • Manufactured to ISO 9001 quality standards
  • Supplied as a two-pack
  • Features a ceramic ball and seat
  • Compatible with HPLC systems