Learn More
Waters Corp Cartridge Check Valve, HPLC Compatible, Two-Piece Housing
Catalog No. 50819006 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The PerformancePLUS Cartridge Check Valve System serves as an inlet or outlet valve in compatible HPLC pumps. Its check valve materials are selected for compatibility with all HPLC mobile phases. The housing features a two-piece design, enabling cartridge replacement without disturbing the fitting.
  • Can be used as either an inlet or outlet valve.
  • Compatible with 1515, 1525, 515, 600, and 610 HPLC pumps.
  • Check valve materials are compatible with all HPLC mobile phases.
  • Two-piece housing design allows cartridge changes without disturbing the fitting.
  • Designed to remove the need for constant cartridge replacement.
  • Strictly tested for use with all HPLC mobile phases.
  • Cartridge can be removed from housing without disturbing other parts for quick changes.
  • Includes two valve cartridges and two housings.
Catalog No. 50-819-006 Supplier Waters Corp Supplier No. 700000253
The PerformancePLUS Cartridge Check Valve System serves as an inlet or outlet valve in compatible HPLC pumps. Its check valve materials are selected for compatibility with all HPLC mobile phases. The housing features a two-piece design, enabling cartridge replacement without disturbing the fitting.
  • Can be used as either an inlet or outlet valve.
  • Compatible with 1515, 1525, 515, 600, and 610 HPLC pumps.
  • Check valve materials are compatible with all HPLC mobile phases.
  • Two-piece housing design allows cartridge changes without disturbing the fitting.
  • Designed to remove the need for constant cartridge replacement.
  • Strictly tested for use with all HPLC mobile phases.
  • Cartridge can be removed from housing without disturbing other parts for quick changes.
  • Includes two valve cartridges and two housings.