Learn More
Waters Corp Rebuild Kit, PerformancePLUS Alliance Seal Pack, HPLC
Catalog No. 502392608 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit contains a replacement injector needle and the necessary seals, spacers and frits necessary to rebuild a PerformancePLUS Alliance Seal Pack.
  • Type: Performance maintenance, rebuild kit
  • System type: HPLC
  • Replacement schedule: Annual
  • Component: 2695 Separations Module
  • Suitable for 2695s manufactured after March 2018 or updated with enhancement kit 700011805
  • Compatible with Alliance HPLC System
Catalog No. 50-239-2608 Supplier Waters Corp Supplier No. 700011783
This rebuild kit contains a replacement injector needle and the necessary seals, spacers and frits necessary to rebuild a PerformancePLUS Alliance Seal Pack.
  • Type: Performance maintenance, rebuild kit
  • System type: HPLC
  • Replacement schedule: Annual
  • Component: 2695 Separations Module
  • Suitable for 2695s manufactured after March 2018 or updated with enhancement kit 700011805
  • Compatible with Alliance HPLC System