Learn More
Waters Corp Performance PLUS Seal Pack Replacement Kit, Alliance HPLC System, 2695 Separations Module
Catalog No. 502392861 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains a replacement injector needle, a restrictor loop, and a complete PerformancePLUS Seal Pack Assembly. It is designed for use with Alliance HPLC Systems, particularly for the maintenance of 2695 Separations Modules.
  • Contains a replacement injector needle, restrictor loop, and a complete PerformancePLUS Seal Pack Assembly.
  • Designed for use with HPLC systems.
  • Compatible with 2695 Separations Modules.
  • Suitable for modules manufactured after March 2018 or those updated with an enhancement kit.
  • Replaces the injector seal pack assembly.
Catalog No. 50-239-2861 Supplier Waters Corp Supplier No. 700011784
This kit contains a replacement injector needle, a restrictor loop, and a complete PerformancePLUS Seal Pack Assembly. It is designed for use with Alliance HPLC Systems, particularly for the maintenance of 2695 Separations Modules.
  • Contains a replacement injector needle, restrictor loop, and a complete PerformancePLUS Seal Pack Assembly.
  • Designed for use with HPLC systems.
  • Compatible with 2695 Separations Modules.
  • Suitable for modules manufactured after March 2018 or those updated with an enhancement kit.
  • Replaces the injector seal pack assembly.