Learn More
Waters Corp Waters Corporation Check Valve Housing, PerformancePLUS, HPLC System, Stainless Steel, 2-Pack
Catalog No. 50818921 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This check valve housing is a replacement cartridge designed for an HPLC system. It is supplied as a 2 pk and is compatible with 2695 and 2695 D Separations Modules.
  • Made from stainless steel
  • Designed for HPLC system
  • Compatible with 2695 Separations Module and 2695 D Dissolution Separations Module
  • Functions as a check valve
  • Supplied as 2 pk
  • Replacement as required
Catalog No. 50-818-921 Supplier Waters Corp Supplier No. 700002332
This check valve housing is a replacement cartridge designed for an HPLC system. It is supplied as a 2 pk and is compatible with 2695 and 2695 D Separations Modules.
  • Made from stainless steel
  • Designed for HPLC system
  • Compatible with 2695 Separations Module and 2695 D Dissolution Separations Module
  • Functions as a check valve
  • Supplied as 2 pk
  • Replacement as required