Learn More
Thermco Products PerformaTherm™ PCM Panels
Catalog No. 04500328
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
04-500-328 Non-sterile
1 options
Catalog No. 04-500-328 Supplier Thermco Products Supplier No. ACCPTPC
Add to Cart
Add to Cart
PerformaTherm™ PCM Panels, Set of 6.
PerformaTherm™ Phase Change Material (PCM) when frozen will maintain stable temperature (2°C to 8°C) when inserted into a Performa Therm Medical Transport Carrier.
Specifications
Product Type PCM Panels
Sterility Non-sterile