Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermco Products PerformaTherm™ Validated Medical Transport Carrier

Catalog No. 04500360
Change view
Click to view available options
Volume (Metric):
3 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
04-500-360 3 L
04-500-322 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-500-360 Supplier Thermco Products Supplier No. ACCPT3L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Performa-Therm validated medical transport carrier is ISTA 7E Validated. Zipper closure with carry handles and shoulder strap.

  • The Performa Therm portable validated medical transport carrier maintains product integrity, ensures compliance, and delivers life-saving treatments with precise temperature control.
  • Ideal for short-term storage, carrying, and delivery of temperature-sensitive materials such as tissue, serum, vaccines, cultures, specimens, and other items requiring 2°C to 8°C conditions.
  • Features a durable, reusable, and washable nylon shell with a molded tub and six PCM panels.
  • ISTA 7E validated to maintain temperatures between 2°C and 8°C for up to 48 hr.
  • Supplied with a validation certificate.

Specifications

Product Type Medical Transport Tote
Sterility Non-sterile
Volume (Metric) 3 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.