Learn More
Waters Corp Waters Autosampler Needle, MP35N, Corrosion Resistant, HPLC
Catalog No. 50819029 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The PerformancePLUS Autosampler Needle is constructed from corrosion-resistant nickel/cobalt alloy (MP35N). This design makes the needles less susceptible to etching by acids, potentially resulting in reduced protein absorption and lower carryover compared to standard stainless steel needles when used in acidic solutions. This specific needle assembly is designed for use with the 717 Plus autosampler.
  • Constructed from corrosion-resistant nickel/cobalt alloy (MP35N).
  • Significantly reduced susceptibility to etching by acids.
  • Lower protein absorption and reduced carryover compared to standard stainless steel needles when used in acidic solutions.
  • Designed for use with the 717 Plus autosampler.
  • Needle type.
  • HPLC system compatible.
  • 1 unit per package.
  • Annual replacement schedule.
Catalog No. 50-819-029 Supplier Waters Corp Supplier No. 700001247
The PerformancePLUS Autosampler Needle is constructed from corrosion-resistant nickel/cobalt alloy (MP35N). This design makes the needles less susceptible to etching by acids, potentially resulting in reduced protein absorption and lower carryover compared to standard stainless steel needles when used in acidic solutions. This specific needle assembly is designed for use with the 717 Plus autosampler.
  • Constructed from corrosion-resistant nickel/cobalt alloy (MP35N).
  • Significantly reduced susceptibility to etching by acids.
  • Lower protein absorption and reduced carryover compared to standard stainless steel needles when used in acidic solutions.
  • Designed for use with the 717 Plus autosampler.
  • Needle type.
  • HPLC system compatible.
  • 1 unit per package.
  • Annual replacement schedule.