Learn More
Electron Microscopy Sciences Perf-Tape™ Perforated Tape 3/4", Blue 2160 IN
SDP
Catalog No. 501926803
Encompass_Preferred
Slit-perforated laboratory tape eliminates the need for scissors. Slit-perforated every 1.25" to allow for clean tearing every time. Quick tear and apply for use. High-tech perforations are designed to remain "invisible" when tape is marked and may be written over. Perf-Tape won't tear while it is being unrolled. Leaves no adhesive residue when peeled off. Water resistant and also oil and acid resistant. Withstands cold (-23 degree C) and hot (121 degree C) temperatures.
Color: Blue
Catalog No. 50-192-6803 Supplier Electron Microscopy Sciences Supplier No. 3677B
Slit-perforated laboratory tape eliminates the need for scissors. Slit-perforated every 1.25" to allow for clean tearing every time. Quick tear and apply for use. High-tech perforations are designed to remain "invisible" when tape is marked and may be written over. Perf-Tape won't tear while it is being unrolled. Leaves no adhesive residue when peeled off. Water resistant and also oil and acid resistant. Withstands cold (-23 degree C) and hot (121 degree C) temperatures.
Color: Blue