Learn More
Electron Microscopy Sciences CoverWell™ Perfusion Chamber 3 well with 120 µl volume
SDP
Catalog No. 5027776
Encompass_Preferred
CoverWell Perfusion Chambers allows up to 16 different specimens on one coverglass. "Press-to-Seal" covers form water-tight, multiwall cell incubation or cytochemistry chamber when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports (1.5 mm Dia) without disturbing or cross-contaminating specimens in adjacent wells. Chambers are removable for permanent mounting of specimens. CoverWell chambers are made from medical grade silicone bonded to clear, UV transparent plastic.
Volume: 120 µl
Catalog No. 50-277-76 Supplier Electron Microscopy Sciences Supplier No. 7032632
CoverWell Perfusion Chambers allows up to 16 different specimens on one coverglass. "Press-to-Seal" covers form water-tight, multiwall cell incubation or cytochemistry chamber when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports (1.5 mm Dia) without disturbing or cross-contaminating specimens in adjacent wells. Chambers are removable for permanent mounting of specimens. CoverWell chambers are made from medical grade silicone bonded to clear, UV transparent plastic.
Volume: 120 µl