Learn More
Electron Microscopy Sciences Perfusion Fixative, Super Reagent Grade, pH 1000 ML
SDP
Catalog No. 5033442
Encompass_Preferred
For all of the best results we offer ready to use Perfusion washes and fixatives which are recommended by the NeuroScience Association. All of our solutions are ready made and are of the highest quality. Perfusion wash and fixative superior reagent grade. For best results. 4% Paraformaldehyde/4% Sucrose/ 1.4% Sodium Cacodylate.
Grade: Super Reagent
Catalog No. 50-334-42 Supplier Electron Microscopy Sciences Supplier No. 1223SK
For all of the best results we offer ready to use Perfusion washes and fixatives which are recommended by the NeuroScience Association. All of our solutions are ready made and are of the highest quality. Perfusion wash and fixative superior reagent grade. For best results. 4% Paraformaldehyde/4% Sucrose/ 1.4% Sodium Cacodylate.
Grade: Super Reagent