Learn More
Ibidi USA Perfusion Set, Red
SDP
Catalog No. NC1779128
Encompass
Ready to use tubing sets for flow assays with the ibidi Pump System. Includes tubing and resvoirs to perform flow assays in combination with the ibidi Channel Slides and Fluidic Units. Product Variation: Red; length 15 cm, ID 1.6 mm, 10 ml reservoirsPcs./Box: 4 (individually packed)
This item is not returnable. View return policy
Catalog No. NC1779128 Supplier Ibidi USA Supplier No. 10962
Ready to use tubing sets for flow assays with the ibidi Pump System. Includes tubing and resvoirs to perform flow assays in combination with the ibidi Channel Slides and Fluidic Units. Product Variation: Red; length 15 cm, ID 1.6 mm, 10 ml reservoirsPcs./Box: 4 (individually packed)