Learn More
Ibidi USA Perfusion Set, Yellow/Green
SDP
Catalog No. NC2020036
Encompass
Ready to use tubing sets for flow assays with the ibidi Pump System. Includes tubing and resvoirs to perform flow assays in combination with the ibidi Channel Slides and Fluidic Units. Product Variation: YELLOW-GREEN; length 50 cm, ID 1.6 mm, 10 ml reservoirsPcs./Box: 4 (individually packed)
This item is not returnable. View return policy
Catalog No. NC2020036 Supplier Ibidi USA Supplier No. 10964
Ready to use tubing sets for flow assays with the ibidi Pump System. Includes tubing and resvoirs to perform flow assays in combination with the ibidi Channel Slides and Fluidic Units. Product Variation: YELLOW-GREEN; length 50 cm, ID 1.6 mm, 10 ml reservoirsPcs./Box: 4 (individually packed)