Learn More
Electron Microscopy Sciences Perfusion Wash, Reagent Grade, 7.2-7.4 pH 1000 ML
SDP
Catalog No. 5033438
Encompass_Preferred
For all of the best results we offer ready to use Perfusion washes and fixatives which are recommended by the NeuroScience Association. All of our solutions are ready made and are of the highest quality. Perfusion wash and fixative reagent grade. To be used for Standard and ImmunoHistochemistry Stains. Phosphate Buffered Saline 0.1M Phosphate Buffer pH 7.2-7.4.
Grade: Reagent
Catalog No. 50-334-38 Supplier Electron Microscopy Sciences Supplier No. 1219SK
For all of the best results we offer ready to use Perfusion washes and fixatives which are recommended by the NeuroScience Association. All of our solutions are ready made and are of the highest quality. Perfusion wash and fixative reagent grade. To be used for Standard and ImmunoHistochemistry Stains. Phosphate Buffered Saline 0.1M Phosphate Buffer pH 7.2-7.4.
Grade: Reagent