Learn More
Perkin Elmer US LLC pergo 1 Gas Line Conversion Kit for NexION 1000/2000
Catalog No. 502111550
Encompass_Preferred
The pergo improves performance of all concentric nebulizers for the routine analysis of brines, fusion digests and other high-TDS samples.
Catalog No. 50-211-1550 Supplier Perkin Elmer US LLC Supplier No. N8152683
The pergo improves performance of all concentric nebulizers for the routine analysis of brines, fusion digests and other high-TDS samples.