Learn More
Perkin Elmer US LLC pergo 1 Humidifier with PFA-ST Nebulizer
Catalog No. 502112263
Encompass_Preferred
This assembly includes a PFA-ST Nebulizer for enhanced aspiration. The PFA-ST nebulizer features high purity, HF-resistance and high performance, along with an exchangeable external sample uptake capillary. It is chemically resistant and ideal for strong acids, alkalis, and organics. This assembly includes: pergo Heating Unit PFA Water Reservoir pergo Power Supply Kit -12V Power Supply, user replaceable adapter plugs for North America/Japan, United Kingdom, Europe, and Australia Gas Line Conversion Kit Gas Line Nebulizer Gas Line Sample Transfer Line for MEINHARD Plus and PFA-ST Nebulizers Gas Fitting Connectors, Pkg. 10 PFA-ST Nebulizer
Catalog No. 50-211-2263 Supplier Perkin Elmer US LLC Supplier No. N8150427
Add to Cart
Add to Cart
Request a Quote
This assembly includes a PFA-ST Nebulizer for enhanced aspiration. The PFA-ST nebulizer features high purity, HF-resistance and high performance, along with an exchangeable external sample uptake capillary. It is chemically resistant and ideal for strong acids, alkalis, and organics. This assembly includes: pergo Heating Unit PFA Water Reservoir pergo Power Supply Kit -12V Power Supply, user replaceable adapter plugs for North America/Japan, United Kingdom, Europe, and Australia Gas Line Conversion Kit Gas Line Nebulizer Gas Line Sample Transfer Line for MEINHARD Plus and PFA-ST Nebulizers Gas Fitting Connectors, Pkg. 10 PFA-ST Nebulizer