Learn More
Perkin Elmer US LLC pergo 2 Humidifier with MEINHARD Plus Type C Nebulizer
Supplier:  Perkin Elmer US LLC N8150428
Encompass_Preferred
This assembly includes a Type C MEINHARD Plus High Solids Nebulizer for enhanced aspiration. The MEINHARD Concentric Nebulizer reflects exacting standards in design and manufacturing. Its advantages include simplicity of design and operation, reproducible and self-aligning aerosol and close fabrication control. They provide excellent sensitivity and precision for aqueous solutions and samples with few dissolved solids (less than 1%). This assembly includes: pergo Heating Unit PFA Water Reservoir pergo Power Supply Kit -12V Power Supply, user replaceable adapter plugs for North America/Japan, United Kingdom, Europe, and Australia Gas Line Conversion Kit Gas Line Nebulizer Gas Line Sample Transfer Line for MEINHARD Plus and PFA-ST Nebulizers Gas Fitting Connectors, Pkg. 10 Plus Series glass concentric nebulizer, Type C, high solids. 1 L/min and 0.5 mL/min at 50 psi
Catalog No. 50-211-2259