Learn More
Perkin Elmer US LLC pergo 2 Humidifier without Nebulizer
Catalog No. 502113287
Encompass_Preferred
This assembly includes: pergo main unit (heater), Qty. 1 PFA water reservoir with min/max lines and four ported caps, Qty. 1 pergo spare power supply, 12V, 1.5A, wall mount with plugs for North America, Europe, UK, Qty. 1 2.4 mm I.D. x 4mm O.D. PTFE gas line for MicroFlow nebulizer, 1 m, Qty. 4 Gas fitting, Pkg. 10 Note: This assembly does not include a nebulizer.
Catalog No. 50-211-3287 Supplier Perkin Elmer US LLC Supplier No. N8140743
This assembly includes: pergo main unit (heater), Qty. 1 PFA water reservoir with min/max lines and four ported caps, Qty. 1 pergo spare power supply, 12V, 1.5A, wall mount with plugs for North America, Europe, UK, Qty. 1 2.4 mm I.D. x 4mm O.D. PTFE gas line for MicroFlow nebulizer, 1 m, Qty. 4 Gas fitting, Pkg. 10 Note: This assembly does not include a nebulizer.