Learn More
Apexbio Technology LLC Pergolide mesylate 66104-23-2 25mg
SDP
Supplier:  Apexbio Technology LLC B148525
Encompass_Preferred
Pergolide mesylate is a small-molecule agonist targeting dopamine D2-family receptors It is designed to activate these receptors thereby modulating dopaminergic pathways in the central nervous system Pergolide mesylate exerts its biological activity primarily through agonism of dopamine receptors Based on these pharmacological properties pergolide mesylate holds research potential in studies of dopaminergic neurotransmission receptor signaling pathways Parkinson s disease mechanisms and the investigation of therapeutic mechanisms for neurodegenerative disorders characterized by dopaminergic deficits
Catalog No. 50-000-89967