Learn More
Medchemexpress LLC Pergolide (mesylate) | 66104-23-2 | 410.59 | 50 MG
SDP
Catalog No. 5000348533
Encompass_Preferred
Pergolide mesylate, an Ergoline derivative, is a potent and orally active dopamine D1 and D2 receptors agonist. It can be used for Parkinson's disease and hyperprolactinaemia research.
  • Potent and orally active dopamine D1 and D2 receptors agonist.
  • Inhibits H2O2-induced cell death in SH-SY5Y neuroblastoma cells.
  • Protects SH-SY5Y neuroblastoma cells from cell death at low concentrations.
  • Reduces working/reference memory errors in a rat model of Parkinson's disease.
  • Facilitates spatial memory and improves brain oxidative balance in animal models.
Catalog No. 50-003-48533 Supplier Medchemexpress LLC Supplier No. HY13720A50MG
Pergolide mesylate, an Ergoline derivative, is a potent and orally active dopamine D1 and D2 receptors agonist. It can be used for Parkinson's disease and hyperprolactinaemia research.
  • Potent and orally active dopamine D1 and D2 receptors agonist.
  • Inhibits H2O2-induced cell death in SH-SY5Y neuroblastoma cells.
  • Protects SH-SY5Y neuroblastoma cells from cell death at low concentrations.
  • Reduces working/reference memory errors in a rat model of Parkinson's disease.
  • Facilitates spatial memory and improves brain oxidative balance in animal models.