Learn More
Medchemexpress LLC Pergolide mesylate | 66104-23-2 | 99.98% | 410.59 | 25 MG
SDP
Catalog No. 5000351500
Encompass_Preferred
Pergolide mesylate (Pergolide methanesulfonate) is an Ergoline derivative, and a potent and orally active dopamine D1 and D2 receptors agonist. It can be utilized for research into Parkinson's disease and hyperprolactinaemia.
  • Potent and orally active
  • Dopamine D1 and D2 receptors agonist
  • Utilized for research into Parkinson's disease
  • Utilized for research into hyperprolactinaemia
Catalog No. 50-003-51500 Supplier Medchemexpress LLC Supplier No. 25MGHY13720A
Pergolide mesylate (Pergolide methanesulfonate) is an Ergoline derivative, and a potent and orally active dopamine D1 and D2 receptors agonist. It can be utilized for research into Parkinson's disease and hyperprolactinaemia.
  • Potent and orally active
  • Dopamine D1 and D2 receptors agonist
  • Utilized for research into Parkinson's disease
  • Utilized for research into hyperprolactinaemia