Learn More
Medchemexpress LLC Pergolide mesylate | 66104-23-2 | 99.98% | 410.59 | 5 MG
SDP
Catalog No. 5000364351
Encompass_Preferred
Pergolide mesylate (Pergolide methanesulfonate) is an Ergoline derivative, a potent and orally active dopamine D1 and D2 receptors agonist. It is used in research for Parkinson's disease and hyperprolactinaemia.
  • Potent and orally active dopamine D1 and D2 receptors agonist.
  • Used in research for Parkinson's disease.
  • Used in research for hyperprolactinaemia.
  • Inhibits H2O2-induced cell death in SH-SY5Y neuroblastoma cells.
  • Protects SH-SY5Y neuroblastoma cells from H2O2-induced cell death at nanomolar concentrations.
  • Reduces working/reference memory errors in a rat model of Parkinson's disease.
  • Facilitates spatial memory and improves brain oxidative balance in a rat model of Parkinson's disease.
Catalog No. 50-003-64351 Supplier Medchemexpress LLC Supplier No. 5MGHY13720A
Pergolide mesylate (Pergolide methanesulfonate) is an Ergoline derivative, a potent and orally active dopamine D1 and D2 receptors agonist. It is used in research for Parkinson's disease and hyperprolactinaemia.
  • Potent and orally active dopamine D1 and D2 receptors agonist.
  • Used in research for Parkinson's disease.
  • Used in research for hyperprolactinaemia.
  • Inhibits H2O2-induced cell death in SH-SY5Y neuroblastoma cells.
  • Protects SH-SY5Y neuroblastoma cells from H2O2-induced cell death at nanomolar concentrations.
  • Reduces working/reference memory errors in a rat model of Parkinson's disease.
  • Facilitates spatial memory and improves brain oxidative balance in a rat model of Parkinson's disease.