Learn More
Medchemexpress LLC Pergolide (mesylate) | 66104-23-2 | HY-13720A | 100.0% | 410.59 | 10 MG
SDP
Catalog No. 5000365329
Encompass_Preferred
Pergolide mesylate (Pergolide methanesulfonate) is an Ergoline derivative that acts as a potent and orally active agonist for dopamine D1 and D2 receptors. It can be used for Parkinson's disease and hyperprolactinaemia research.
  • Acts as a potent and orally active agonist for dopamine D1 and D2 receptors.
  • Used in research for Parkinson's disease.
  • Used in research for hyperprolactinaemia.
  • Inhibits H2O2-induced cell death in SH-SY5Y neuroblastoma cells.
  • Protects SH-SY5Y neuroblastoma cells from cell death.
  • Reduces working/reference memory errors in a rat model of Parkinson's disease.
  • Facilitates spatial memory.
  • Improves brain oxidative balance.
Catalog No. 50-003-65329 Supplier Medchemexpress LLC Supplier No. 10MGHY13720A
Pergolide mesylate (Pergolide methanesulfonate) is an Ergoline derivative that acts as a potent and orally active agonist for dopamine D1 and D2 receptors. It can be used for Parkinson's disease and hyperprolactinaemia research.
  • Acts as a potent and orally active agonist for dopamine D1 and D2 receptors.
  • Used in research for Parkinson's disease.
  • Used in research for hyperprolactinaemia.
  • Inhibits H2O2-induced cell death in SH-SY5Y neuroblastoma cells.
  • Protects SH-SY5Y neuroblastoma cells from cell death.
  • Reduces working/reference memory errors in a rat model of Parkinson's disease.
  • Facilitates spatial memory.
  • Improves brain oxidative balance.