Learn More
Cayman Chemical PergolIdemesylate 250mg
SDP
Catalog No. 502934327
Encompass_Preferred
A potent and selective Syk inhibitor (IC50 4 nM) inhibits 92 of Syk activity while other kinases retain 70 activity at a concentration of 50 nM in a panel of 270 kinases inhibits convulxin-induced aggregation of human platelet-rich plasma (IC50 2.5 M) prevents thrombosis in a transgenic mouse model of heparin-induced thrombocytopenia induces CLL B cell apoptosis and inhibits the secretion of chemokines CCL3 CCL4 and CXCL13 inhibits CLL B cell chemotaxis and pseudoemperipolesis
Catalog No. 50-293-4327 Supplier Cayman Chemical Supplier No. 26085250 MG
A potent and selective Syk inhibitor (IC50 4 nM) inhibits 92 of Syk activity while other kinases retain 70 activity at a concentration of 50 nM in a panel of 270 kinases inhibits convulxin-induced aggregation of human platelet-rich plasma (IC50 2.5 M) prevents thrombosis in a transgenic mouse model of heparin-induced thrombocytopenia induces CLL B cell apoptosis and inhibits the secretion of chemokines CCL3 CCL4 and CXCL13 inhibits CLL B cell chemotaxis and pseudoemperipolesis