Learn More
Cayman Chemical Pergolide (mesylate), Purity: ≥95%, CAS: 66104-23-2, 50 mg
SDP
Catalog No. 502936265
Encompass_Preferred
A dopamine D1 and D2 receptor agonist (Kis = 111 and 0.495 nM, respectively, for rat striatal receptors); depresses dopaminergic firing in paralyzed rats (ED50 = 50 = 0.4 mg/kg)
Catalog No. 50-293-6265 Supplier Cayman Chemical Supplier No. 2608550 MG
A dopamine D1 and D2 receptor agonist (Kis = 111 and 0.495 nM, respectively, for rat striatal receptors); depresses dopaminergic firing in paralyzed rats (ED50 = 50 = 0.4 mg/kg)