Learn More
TARGETMOL CHEMICALS INC PERHEXILINE MALEATE 10MG
SDP
Catalog No. 502577424
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Perhexiline maleate is an orally active inhibitor of CPT1 and CPT2 that reduces fatty acid metabolism. The IC50 values of perhexiline maleate against rat heart and liver CPT 1 were 77 and 148 uM respectively. purity: 99%
Catalog No. 50-257-7424 Supplier TARGETMOL CHEMICALS INC Supplier No. T1240810MG
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Perhexiline maleate is an orally active inhibitor of CPT1 and CPT2 that reduces fatty acid metabolism. The IC50 values of perhexiline maleate against rat heart and liver CPT 1 were 77 and 148 uM respectively. purity: 99%