Learn More
Selleck Chemical LLC Perhexiline maleate
SDP
Catalog No. 5000002498
Encompass_Preferred
Perhexiline maleate is a potent inhibitor of carnitine palmitoyltransferase 1 (CPT1) with IC50s of 77 M and 148 M for rat heart CPT1 and liver CPT1 respectively and also inhibits carnitine palmitoyltransferase 2 (CPT2)
Catalog No. 50-000-02498 Supplier Selleck Chemical LLC Supplier No. S6959100MG
Perhexiline maleate is a potent inhibitor of carnitine palmitoyltransferase 1 (CPT1) with IC50s of 77 M and 148 M for rat heart CPT1 and liver CPT1 respectively and also inhibits carnitine palmitoyltransferase 2 (CPT2)