Learn More
Selleck Chemical LLC Perhexiline maleate 5mg
SDP
Supplier:  Selleck Chemical LLC S69595MG
Encompass_Preferred
Perhexiline maleate is a potent inhibitor of carnitine palmitoyltransferase 1 (CPT1) with IC50 of 77 μM and 148 μM for rat heart CPT1 and liver CPT1, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3241