Learn More
Medchemexpress LLC Perhexiline (maleate) | 6724-53-4 | 100.0% | 1 ML
SDP
Catalog No. 5000412005
Encompass_Preferred
Perhexiline maleate is an orally active CPT1 and CPT2 inhibitor that reduces fatty acid metabolism. It induces mitochondrial dysfunction and apoptosis in hepatic cells. It can cross the blood-brain barrier (BBB) and exhibits anti-tumor activity. Perhexiline maleate is used in the research of cancers and cardiovascular diseases like angina.
  • Orally active CPT1 and CPT2 inhibitor
  • Reduces fatty acid metabolism
  • Induces mitochondrial dysfunction and apoptosis in hepatic cells
  • Can cross the blood-brain barrier (BBB)
  • Shows anti-tumor activity
  • Useful in research for cancers and cardiovascular diseases like angina
Catalog No. 50-004-12005 Supplier Medchemexpress LLC Supplier No. HYB1334A10MM/1ML
Perhexiline maleate is an orally active CPT1 and CPT2 inhibitor that reduces fatty acid metabolism. It induces mitochondrial dysfunction and apoptosis in hepatic cells. It can cross the blood-brain barrier (BBB) and exhibits anti-tumor activity. Perhexiline maleate is used in the research of cancers and cardiovascular diseases like angina.
  • Orally active CPT1 and CPT2 inhibitor
  • Reduces fatty acid metabolism
  • Induces mitochondrial dysfunction and apoptosis in hepatic cells
  • Can cross the blood-brain barrier (BBB)
  • Shows anti-tumor activity
  • Useful in research for cancers and cardiovascular diseases like angina