Learn More
Medchemexpress LLC Perhexiline (maleate) | 6724-53-4 | 100 MG
SDP
Catalog No. 5000412100
Encompass_Preferred
Perhexiline maleate is an orally active CPT1 and CPT2 inhibitor that reduces fatty acid metabolism. It induces mitochondrial dysfunction and apoptosis in hepatic cells. It can cross the blood brain barrier (BBB) and shows anti-tumor activity. It can be used in the research of cancers, and cardiovascular diseases like angina.
  • Reduces fatty acid metabolism
  • Induces mitochondrial dysfunction and apoptosis in hepatic cells
  • Can cross the blood-brain barrier
  • Exhibits anti-tumor activity
  • Suitable for research in cancers and cardiovascular diseases like angina
Catalog No. 50-004-12100 Supplier Medchemexpress LLC Supplier No. HYB1334A100MG
Perhexiline maleate is an orally active CPT1 and CPT2 inhibitor that reduces fatty acid metabolism. It induces mitochondrial dysfunction and apoptosis in hepatic cells. It can cross the blood brain barrier (BBB) and shows anti-tumor activity. It can be used in the research of cancers, and cardiovascular diseases like angina.
  • Reduces fatty acid metabolism
  • Induces mitochondrial dysfunction and apoptosis in hepatic cells
  • Can cross the blood-brain barrier
  • Exhibits anti-tumor activity
  • Suitable for research in cancers and cardiovascular diseases like angina