Learn More
Medchemexpress LLC Perhexiline maleate | 6724-53-4 | 99.97% | 200 MG
SDP
Catalog No. 5000411979
Encompass_Preferred
Perhexiline maleate is an orally active inhibitor of CPT1 and CPT2, which functions to reduce fatty acid metabolism. This compound is known to induce mitochondrial dysfunction and apoptosis in hepatic cells. It can cross the blood-brain barrier and has demonstrated anti-tumor activity, making it relevant for research in cancers and cardiovascular diseases such as angina.
  • Reduces fatty acid metabolism.
  • Induces mitochondrial dysfunction in hepatic cells.
  • Induces apoptosis in hepatic cells.
  • Crosses the blood-brain barrier.
  • Shows anti-tumor activity.
  • Used in research for cancers and cardiovascular disease like angina.
Catalog No. 50-004-11979 Supplier Medchemexpress LLC Supplier No. HYB1334A200MG
Perhexiline maleate is an orally active inhibitor of CPT1 and CPT2, which functions to reduce fatty acid metabolism. This compound is known to induce mitochondrial dysfunction and apoptosis in hepatic cells. It can cross the blood-brain barrier and has demonstrated anti-tumor activity, making it relevant for research in cancers and cardiovascular diseases such as angina.
  • Reduces fatty acid metabolism.
  • Induces mitochondrial dysfunction in hepatic cells.
  • Induces apoptosis in hepatic cells.
  • Crosses the blood-brain barrier.
  • Shows anti-tumor activity.
  • Used in research for cancers and cardiovascular disease like angina.