Learn More
Medchemexpress LLC Perhexiline maleate | 6724-53-4 | MFCD00057329 | 98.0% | 393.56 | C23H39NO4 | 10 MG
SDP
Catalog No. 5000297795
Encompass_Preferred
Perhexiline maleate is the maleate salt of perhexiline, a small-molecule inhibitor of carnitine palmitoyltransferase 1 and 2 (CPT1/CPT2) used in preclinical research to probe mitochondrial fatty-acid metabolism, cancer biology, and cardiovascular disease models. It is provided as a solid salt intended for biochemical and cell-based assays.
  • Inhibits CPT1 and CPT2, reducing fatty-acid oxidation
  • Maleate salt form improves aqueous handling for assays
  • Available in research-scale quantities, including 10 mg
  • High reported purity suitable for in vitro and preclinical studies
  • Documented chemical identifiers include CAS 6724-53-4 and molecular weight 393.56
Catalog No. 50-002-97795 Supplier Medchemexpress LLC Supplier No. HYB1334A10MG
Perhexiline maleate is the maleate salt of perhexiline, a small-molecule inhibitor of carnitine palmitoyltransferase 1 and 2 (CPT1/CPT2) used in preclinical research to probe mitochondrial fatty-acid metabolism, cancer biology, and cardiovascular disease models. It is provided as a solid salt intended for biochemical and cell-based assays.
  • Inhibits CPT1 and CPT2, reducing fatty-acid oxidation
  • Maleate salt form improves aqueous handling for assays
  • Available in research-scale quantities, including 10 mg
  • High reported purity suitable for in vitro and preclinical studies
  • Documented chemical identifiers include CAS 6724-53-4 and molecular weight 393.56