Learn More
Sigma Aldrich Fine Chemicals Biosciences Perhexiline maleate salt 950MG
Catalog No. 502886457
Encompass
Perhexiline maleate regulatesnbspcoronary vasodilatation and blocks exercise-induced tachycardia. It is used to treat angina pectoris and variant angina. Perhexiline maleate functions as a cardiac metabolic agent. It is associated with peripheral neuropathy, high intracranial pressure withnbsppapilledemanbspand proximal myopathy.
Catalog No. 50-288-6457 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SML012050MG
Perhexiline maleate regulatesnbspcoronary vasodilatation and blocks exercise-induced tachycardia. It is used to treat angina pectoris and variant angina. Perhexiline maleate functions as a cardiac metabolic agent. It is associated with peripheral neuropathy, high intracranial pressure withnbsppapilledemanbspand proximal myopathy.