Learn More
Cayman Chemical PerhexIlInmaleate 10mg
SDP
Catalog No. 502968147
Encompass_Preferred
A CPT1 and CPT2 inhibitor that inhibits rat heart and liver CPT1 (IC50 = 77 and 148 µM, respectively), and rat heart CPT2 (IC50 = 79 µM); inhibits mTORC1 signaling and induces autophagy in MCF-7 cells at low µM levels
Catalog No. 50-296-8147 Supplier Cayman Chemical Supplier No. 1698210 MG
A CPT1 and CPT2 inhibitor that inhibits rat heart and liver CPT1 (IC50 = 77 and 148 µM, respectively), and rat heart CPT2 (IC50 = 79 µM); inhibits mTORC1 signaling and induces autophagy in MCF-7 cells at low µM levels