Learn More
OPTI MEDICAL PERI PUMP CATRIDGE
Catalog No. NC0957458
Encompass
  • Peripump cartridge kit designed for sample aspiration and transport.
  • New OEM original component for seamless replacement.
  • Compatible with OPTI Blood Gas Analyzers, including the OPTI CCA-TS2.
  • Critical for automated fluidic management in diagnostic systems.
  • Contributes to maintenance-free operation of analyzers.
  • Supports optical fluorescence technology indirectly through integration with analyzers.
  • 90-day warranty ensures reliability and support.
Catalog No. NC0957458 Supplier OPTI MEDICAL Supplier No. BP7012
  • Peripump cartridge kit designed for sample aspiration and transport.
  • New OEM original component for seamless replacement.
  • Compatible with OPTI Blood Gas Analyzers, including the OPTI CCA-TS2.
  • Critical for automated fluidic management in diagnostic systems.
  • Contributes to maintenance-free operation of analyzers.
  • Supports optical fluorescence technology indirectly through integration with analyzers.
  • 90-day warranty ensures reliability and support.