Learn More
Selleck Chemical LLC Cyproheptadine hydrochloride sesquihydrate S4635-500mg
SDP
Catalog No. 503108905
Encompass_Preferred
Cyproheptadine hydrochloride (Periactin Peritol) is non-selective 5HT2 antagonist with IC50 of 0 6 nM Also a SETD7/9 inhibitor
Catalog No. 50-310-8905 Supplier Selleck Chemical LLC Supplier No. S4635500MG
Cyproheptadine hydrochloride (Periactin Peritol) is non-selective 5HT2 antagonist with IC50 of 0 6 nM Also a SETD7/9 inhibitor