Learn More
Medchemexpress LLC Pericyazine | 2622-26-6 | 98.0% | C21H23N3OS | 10MG
SDP
Catalog No. 5000261796
Encompass_Preferred
Pericyazine is a first-generation antipsychotic and selective D2-dopamine receptor antagonist supplied as a small-molecule research reference standard. It is used in pharmacological and biochemical studies to probe dopaminergic signaling and antipsychotic mechanisms, and is documented to have adrenolytic, anticholinergic, and extrapyramidal effects.
  • First-generation antipsychotic reference standard for research.
  • Selective D2-dopamine receptor antagonist activity.
  • Suitable for pharmacology and receptor binding studies.
  • Available in small analytical pack sizes for assay preparation.
  • Documented physicochemical data to support quality control.
Catalog No. 50-002-61796 Supplier Medchemexpress LLC Supplier No. HY1426310MG
Pericyazine is a first-generation antipsychotic and selective D2-dopamine receptor antagonist supplied as a small-molecule research reference standard. It is used in pharmacological and biochemical studies to probe dopaminergic signaling and antipsychotic mechanisms, and is documented to have adrenolytic, anticholinergic, and extrapyramidal effects.
  • First-generation antipsychotic reference standard for research.
  • Selective D2-dopamine receptor antagonist activity.
  • Suitable for pharmacology and receptor binding studies.
  • Available in small analytical pack sizes for assay preparation.
  • Documented physicochemical data to support quality control.