Learn More
Medchemexpress LLC Pericyazine | 2622-26-6 | 99.86% | C21H23N3OS | 50 MG
SDP
Catalog No. 5000353891
Encompass_Preferred
Pericyazine (Propericiazine) is identified as a first-generation antipsychotic agent. It serves as an adjunct for the short-term management of severe anxiety states and psychosis. This compound functions as a selective D2-dopamine receptor antagonist. It also exhibits adrenolytic, anticholinergic, and extrapyramidal effects.
  • First-generation antipsychotic agent.
  • Used for short-term management of severe anxiety states.
  • Used for short-term management of psychosis.
  • Selective D2-dopamine receptor antagonist.
  • Exhibits adrenolytic effects.
  • Exhibits anticholinergic effects.
  • Exhibits extrapyramidal effects.
Catalog No. 50-003-53891 Supplier Medchemexpress LLC Supplier No. HY1426350MG
Pericyazine (Propericiazine) is identified as a first-generation antipsychotic agent. It serves as an adjunct for the short-term management of severe anxiety states and psychosis. This compound functions as a selective D2-dopamine receptor antagonist. It also exhibits adrenolytic, anticholinergic, and extrapyramidal effects.
  • First-generation antipsychotic agent.
  • Used for short-term management of severe anxiety states.
  • Used for short-term management of psychosis.
  • Selective D2-dopamine receptor antagonist.
  • Exhibits adrenolytic effects.
  • Exhibits anticholinergic effects.
  • Exhibits extrapyramidal effects.